Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
ENST00000325411.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
ENST00000350657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
ENST00000456947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
ENST00000463318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
NM_001099646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
NM_152908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016730.hg.1 |
TC17001249.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000325411.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000350657.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000433844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000463318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000467609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000574220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000574239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_001099646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_001256663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_152908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000325411.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000350657.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000433844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000463318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000467609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000574220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
ENST00000574239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_001099646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_001256663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
NM_152908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700178648.hg.1 |
TC1700010035.hg.1 |
Clariom D TC |