Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016826.hg.1 |
ENST00000266505.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016826.hg.1 |
ENST00000447925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016826.hg.1 |
NM_033123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016826.hg.1 |
TC12001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
ENST00000266505.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
ENST00000435379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
ENST00000447925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
ENST00000539875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
NM_033123.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016828.hg.1 |
TC12001291.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000266505.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000318197.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000535429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000541109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000541966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000543242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000544849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000545129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
NM_033123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
NR_073075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000266505.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000318197.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000535429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000539875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000541109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000541966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000543242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000544849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
ENST00000545129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
NM_033123.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
NR_073075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176100.hg.1 |
TC1200010065.hg.1 |
Clariom D TC |