Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014705.3 | NP_055520.3 | 41 - 42 | 4655 | 69 | |
XM_006716188.2 | XP_006716251.1 | 42 - 43 | 4669 | 69 | ||
XM_006716189.2 | XP_006716252.1 | 42 - 43 | 4669 | 69 | ||
XM_017012819.1 | XP_016868308.1 | 42 - 43 | 5042 | 69 | ||
XM_017012820.1 | XP_016868309.1 | 41 - 42 | 4578 | 69 | ||
XM_017012821.1 | XP_016868310.1 | 42 - 43 | 4596 | 69 | ||
XM_017012822.1 | XP_016868311.1 | 42 - 43 | 5187 | 69 | ||
XM_017012823.1 | XP_016868312.1 | 42 - 43 | 4596 | 69 | ||
XM_017012824.1 | XP_016868313.1 | 41 - 42 | 4569 | 69 | ||
XM_017012825.1 | XP_016868314.1 | 42 - 43 | 4596 | 69 | ||
GenBank mRNA | AB018259.2 | - | 26 - 27 | 2768 | 69 | |
AK294722.1 | - | 11 - 12 | 1112 | 69 | ||
AK294820.1 | - | 11 - 12 | 1375 | 69 | ||
AY233380.1 | - | 41 - 42 | 4386 | 69 | ||
BC117688.2 | - | 41 - 42 | 4386 | 40072000 | 69 | |
BC117689.1 | - | 43 - 44 | 4521 | 40072006 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |