Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013440.hg.1 |
ENST00000261383.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013440.hg.1 |
ENST00000396036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013440.hg.1 |
ENST00000415178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013440.hg.1 |
NM_017539.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013440.hg.1 |
TC16000932.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
ENST00000261383.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
ENST00000396036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
NM_017539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
ENST00000261383.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
ENST00000396036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
NM_017539.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174237.hg.1 |
TC1600009671.hg.1 |
Clariom D TC |