Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014237.hg.1 |
ENST00000307257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014237.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014237.hg.1 |
TC11001117.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100170079.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170079.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170079.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170079.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170079.hg.1 |
TC1100009335.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000227349.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000531316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
ENST00000534362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_024806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009335.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170080.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170080.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170080.hg.1 |
ENST00000307257.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170080.hg.1 |
NM_199124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170080.hg.1 |
TC1100009335.hg.1 |
Clariom D TC |