Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134982.1 | 8 - 9 | 1392 | 93 | ||
GenBank mRNA | AK055088.1 | - | 6 - 7 | 1895 | 93 | |
CD013973.1 | - | 8 - 9 | 1355 | 93 |
Set Membership: |
Amplicon length between 86 and 100 P450 Homo sapiens Probe spans exons |