Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
ENST00000278865.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
ENST00000358152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
ENST00000395032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005979.hg.1 |
TC11000504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000526199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000526199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154658.hg.1 |
TC1100007763.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000278865.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000358152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000395032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000525686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000526199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000526199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000528952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
ENST00000534744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031666.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_001031809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007763.hg.1 |
NM_006138.4 |
Refseq |