Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021072.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021072.hg.1 |
ENST00000428532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021072.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021072.hg.1 |
TC11001829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182819.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182819.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182819.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182819.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182819.hg.1 |
TC1100010981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000300146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
ENST00000531919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010981.hg.1 |
NM_152716.2 |
Refseq |