Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014007.hg.1 |
ENST00000328086.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014007.hg.1 |
ENST00000536768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014007.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014007.hg.1 |
TC16000975.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175332.hg.1 |
TC1600009791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000328086.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000565590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
ENST00000569150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009791.hg.1 |
NM_001012981.4 |
Refseq |