Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043363.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043363.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043363.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213617.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |