Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779409 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779400 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15171961 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842495 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15171955 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779379 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842462 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842417 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779419 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779443 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779425 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779403 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842425 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15171951 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779434 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779446 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779417 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779427 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779413 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779408 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779391 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779346 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842503 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779381 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779442 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779407 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842429 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779385 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779366 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842422 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842466 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779376 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779390 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779414 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779420 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779418 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842497 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842492 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842468 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779350 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842461 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779351 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842414 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842421 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842476 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779369 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779394 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779433 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842424 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779363 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842479 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779396 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779405 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779415 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779375 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779424 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779392 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842413 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779439 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779429 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779449 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842436 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842438 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842474 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779399 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779354 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779388 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842435 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779367 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779374 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842507 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842485 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842512 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842504 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15171950 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779430 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779356 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842486 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842451 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15171952 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779382 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779423 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779387 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779378 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779411 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779365 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00779398 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20842449 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
174676 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
AF346606.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
AK024627.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
AK225131.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
BC023588.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
CR602154.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
L08647.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
118255 |
NM_032193.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
2470 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
25270 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P354288 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P354288 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
2098 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P200051 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P128969 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P128969 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
49410 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P30458462 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P354288 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P354288 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171952 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171955 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779375 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779381 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842417 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779376 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171950 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779368 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779392 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779390 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779366 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842421 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842413 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842429 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779379 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779374 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842451 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842436 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779378 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842425 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779369 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779354 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779363 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842438 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779388 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842435 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779391 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779365 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779346 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779367 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779350 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842422 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842417 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779391 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842422 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779369 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171950 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779378 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779388 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842451 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779392 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779354 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779379 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171955 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779366 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842429 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779375 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779374 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842413 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779390 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842438 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779381 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842425 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779376 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842436 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779363 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779367 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779365 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779346 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779368 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00779350 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842421 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20842435 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15171952 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P354288 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P128969 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200051 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779367 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779392 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779357 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779351 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779382 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779353 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779371 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779368 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779378 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779366 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779359 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779390 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779388 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779365 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779386 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779349 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779369 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779384 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779373 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779362 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779376 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779347 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00779375 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P128969 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200051 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P128969 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P200051 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P354288 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
89772 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
63583 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
39857 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P128969 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P200051 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15171959 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779353 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779398 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779414 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779357 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779425 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779419 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779371 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779369 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779400 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779406 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779366 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779368 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779397 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779420 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779347 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779305 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779433 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779413 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15171963 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779367 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779411 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779431 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779439 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779362 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779392 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779444 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779386 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779409 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779388 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779349 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779390 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779307 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779421 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779423 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779442 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779449 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779359 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779417 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779428 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779395 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779351 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15171953 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779446 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15171962 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779404 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779365 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779382 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779384 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779402 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779378 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779376 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15171946 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779436 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00779375 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P128969 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P200051 |
- |
- |