Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014563.hg.1 |
ENST00000337508.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014563.hg.1 |
ENST00000546074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014563.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014563.hg.1 |
TC12001104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
ENST00000542990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009620.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171887.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171887.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171887.hg.1 |
ENST00000337508.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171887.hg.1 |
NM_031474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171887.hg.1 |
TC1200009620.hg.1 |
Clariom D TC |