Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032444.2 | NP_115820.2 | 13 - 14 | 5361 | 84 | |
XM_011522715.2 | XP_011521017.1 | 13 - 14 | 5372 | 84 | ||
XM_017023775.1 | XP_016879264.1 | 10 - 11 | 4766 | 84 | ||
GenBank mRNA | AB058687.1 | - | 2735 | 84 | ||
AK095411.1 | - | 2 - 3 | 1231 | 84 | ||
BC036335.1 | - | 4 - 5 | 2753 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |