Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317950.1 | NP_001304879.1 | 5 - 6 | 1786 | 159 | |
NM_001317952.1 | NP_001304881.1 | 4 - 5 | 1344 | 159 | ||
NM_030767.5 | NP_110394.3 | 5 - 6 | 1731 | 159 | ||
XM_005252244.2 | XP_005252301.1 | 7 - 8 | 1903 | 159 | ||
XM_005252245.1 | XP_005252302.1 | 6 - 7 | 1761 | 159 | ||
XM_005252247.4 | XP_005252304.1 | 5 - 6 | 1782 | 159 | ||
XM_005252248.2 | XP_005252305.1 | 5 - 6 | 1899 | 159 | ||
XM_006717294.1 | XP_006717357.1 | 5 - 6 | 1715 | 159 | ||
XM_006717295.2 | XP_006717358.1 | 5 - 6 | 1899 | 159 | ||
XM_011519063.2 | XP_011517365.1 | 4 - 5 | 1394 | 159 | ||
XM_011519064.2 | XP_011517366.1 | 5 - 6 | 1899 | 159 | ||
XM_011519065.2 | XP_011517367.2 | 5 - 6 | 1374 | 159 | ||
XM_011519066.2 | XP_011517368.1 | 5 - 6 | 1899 | 159 | ||
XM_017015172.1 | XP_016870661.1 | 6 - 7 | 1501 | 159 | ||
GenBank mRNA | AB075848.1 | - | 5 - 6 | 1904 | 159 | |
AK024431.1 | - | 3 - 4 | 679 | 159 | ||
AK131082.2 | - | 5 - 6 | 1786 | 159 | ||
AK160382.1 | - | 5 - 6 | 1786 | 159 | ||
AL833220.1 | - | 2 - 3 | 208 | 159 | ||
AY703039.1 | - | 6 - 7 | 1386 | 159 | ||
AY703040.1 | - | 4 - 5 | 1344 | 159 | ||
AY703041.1 | - | 6 - 7 | 1100 | 159 | ||
AY703042.1 | - | 4 - 5 | 913 | 159 | ||
AY703043.1 | - | 4 - 5 | 1704 | 159 | ||
AY703044.1 | - | 3 - 4 | 1398 | 159 | ||
BC055285.1 | - | 5 - 6 | 1731 | 6143155 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |