Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004701.hg.1 |
TC01000310.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004701.hg.1 |
ENST00000003912.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004701.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004701.hg.1 |
ENST00000374399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004701.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151283.hg.1 |
TC0100007365.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000003912.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000339255.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000358028.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000374399.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000432012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000475663.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
ENST00000488155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007365.hg.1 |
NM_020448.4 |
Refseq |