Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004824.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004824.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004824.hg.1 |
TC14000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004826.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004826.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004826.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004826.hg.1 |
TC14000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004827.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004827.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004827.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004827.hg.1 |
TC14000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008919.hg.1 |
ENST00000404225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004823.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004823.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004823.hg.1 |
TC14000409.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
ENST00000557319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007500.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152615.hg.1 |
TC1400007500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000556491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152616.hg.1 |
TC1400007500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400152610.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152610.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152610.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152610.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152610.hg.1 |
TC1400007500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
ENST00000261783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
ENST00000557120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
NM_001172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152612.hg.1 |
TC1400007500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008919.hg.1 |
ENST00000404225.1 |
Ensembl |