Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
ENST00000317829.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
ENST00000317837.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
ENST00000542840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011584.hg.1 |
TC11000924.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202245.hg.1 |
TC1100013084.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000317829.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000433060.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000537191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
ENST00000539727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_001301007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013084.hg.1 |
NM_130847.2 |
Refseq |