Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270520.1 | NP_001257449.1 | 15 - 16 | 2111 | 66 | |
GenBank mRNA | AK291790.1 | - | 15 - 16 | 2080 | 66 | |
BC024781.1 | - | 5 - 6 | 752 | 66 | ||
BC038428.1 | - | 15 - 16 | 2103 | 4993504 | 66 | |
BC064999.1 | - | 15 - 16 | 2094 | 5784628 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |