Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282644.1 | NP_001269573.1 | 16 - 17 | 1930 | 65 | |
NM_001282645.1 | NP_001269574.1 | 15 - 16 | 1871 | 65 | ||
NM_001323265.1 | NP_001310194.1 | 16 - 17 | 1987 | 65 | ||
NM_001323266.1 | NP_001310195.1 | 15 - 16 | 1869 | 65 | ||
NM_001323267.1 | NP_001310196.1 | 16 - 17 | 1930 | 65 | ||
NM_006257.4 | NP_006248.1 | 16 - 17 | 1932 | 65 | ||
XM_005252496.4 | XP_005252553.1 | 16 - 17 | 2032 | 65 | ||
XM_005252497.4 | XP_005252554.1 | 16 - 17 | 2034 | 65 | ||
XM_006717465.3 | XP_006717528.1 | 18 - 19 | 4427 | 65 | ||
XM_017016410.1 | XP_016871899.1 | 17 - 18 | 4338 | 65 | ||
XM_017016411.1 | XP_016871900.1 | 17 - 18 | 4366 | 65 | ||
GenBank mRNA | AK293935.1 | - | 15 - 16 | 1809 | 65 | |
AK308099.1 | - | 16 - 17 | 1905 | 65 | ||
AK312411.1 | - | 16 - 17 | 1883 | 65 | ||
BC101465.1 | - | 15 - 16 | 1851 | 8068971 | 65 | |
BC113359.1 | - | 15 - 16 | 1851 | 8322411 | 65 | |
BX647657.1 | - | 15 - 16 | 2017 | 65 | ||
L01087.1 | - | 16 - 17 | 1926 | 65 | ||
L07032.1 | - | 15 - 16 | 1853 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |