Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806407 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806445 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806489 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806490 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806394 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806434 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618220 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806483 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618204 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618162 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618163 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618182 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806431 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806465 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618183 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806449 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806419 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618151 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806417 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806421 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618150 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618221 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806471 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806438 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806410 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806476 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806390 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618164 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806447 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806436 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806458 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806402 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806427 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618179 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618158 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806450 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618219 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618226 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618157 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806472 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806478 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618161 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806487 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15792836 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806456 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806428 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618234 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806454 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618240 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618233 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806488 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806391 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806453 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806464 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806432 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806415 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618222 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806437 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806463 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806470 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806448 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806389 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806433 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618178 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806462 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618210 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806443 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806435 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618175 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806444 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P24806439 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P04618167 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AF258592.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AK001262.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AK001692.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AK001983.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AK024471.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
AK097155.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
BC001375.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
BC003176.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
CR593876.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
CR615933.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
166549 |
NM_018235.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
28749 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
659654 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
446818 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
729653 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
116623 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P19712 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P19712 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
29924 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P139511 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P139511 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
43633 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
49829 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31697558 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31697539 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P15580028 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01443221 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01443214 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01443227 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_14_P139511 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P19712 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P19712 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618166 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618178 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806415 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806419 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618163 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618150 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806390 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806417 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806391 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618155 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806407 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806421 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618175 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806425 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806423 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618161 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806420 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618162 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618157 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806394 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618164 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618164 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806394 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618166 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618161 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806407 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618162 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806423 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806417 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806390 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806420 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806415 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806421 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618157 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618163 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618175 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618150 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806425 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618155 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806391 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806419 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24806393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04618178 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P19712 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P139511 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618156 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618159 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618162 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618155 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618167 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618166 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618161 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618152 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618153 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618157 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P04618169 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P04618164 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15792841 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P04618161 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P04618160 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P04618163 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15792840 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15792837 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15792839 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15792838 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P04618162 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P37551210 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P01443221 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P139511 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P139511 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P19712 |
- |
- |
Custom Human Agilent Array 11k, Chromosome 6, Expression |
2494 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
63915 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
29675 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P139511 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P37551205 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01443221 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P17482253 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P37551210 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15792835 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618150 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618202 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618231 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618186 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618184 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618156 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15792838 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618161 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618198 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618194 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618173 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618228 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618223 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618152 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618213 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618191 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618236 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618193 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618234 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618175 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618229 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618179 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618155 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618206 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618201 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618209 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618239 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618211 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618204 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618162 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618170 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618200 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15792833 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618188 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618219 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618207 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618159 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618166 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04618224 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P139511 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01443221 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P37551210 |
- |
- |