Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008128.hg.1 |
ENST00000377962.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008128.hg.1 |
ENST00000448904.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008128.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008128.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008128.hg.1 |
TC13000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009023.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000377962.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000431550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
ENST00000448904.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
NM_001011705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
NM_007015.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161065.hg.1 |
TC1300009023.hg.1 |
Clariom D TC |