Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009694.hg.1 |
TC05000697.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009694.hg.1 |
ENST00000394894.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009694.hg.1 |
ENST00000508792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05009694.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500160386.hg.1 |
TC0500008777.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000394894.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000502338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000503417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000504621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000508792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000511638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
ENST00000513276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008777.hg.1 |
NM_005733.2 |
Refseq |