Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017552.hg.1 |
ENST00000268489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017552.hg.1 |
ENST00000397992.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017552.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017552.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017552.hg.1 |
TC16001259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
ENST00000584072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600184064.hg.1 |
TC1600010825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000268489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000397992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000558842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000563625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
ENST00000584072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_001164766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010825.hg.1 |
NM_006885.3 |
Refseq |