Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005247.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005247.hg.1 |
ENST00000557970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005247.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005247.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000621767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
ENST00000621767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005246.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005246.hg.1 |
ENST00000557970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005246.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005246.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005249.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005249.hg.1 |
ENST00000557970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005249.hg.1 |
ENST00000559572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005249.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005249.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005250.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005250.hg.1 |
ENST00000559572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005250.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005250.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005251.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005251.hg.1 |
ENST00000557970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005251.hg.1 |
ENST00000559572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005251.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005251.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005245.hg.1 |
ENST00000254457.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005245.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005245.hg.1 |
TC17000443.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000621767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
ENST00000621767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007657.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000619297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155213.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155211.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155211.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155211.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155211.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155211.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155214.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155216.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155218.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155219.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155212.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000616237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155220.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
ENST00000614239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
ENST00000619939.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
NM_005568.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155215.hg.1 |
TC1700007657.hg.1 |
Clariom D TC |