Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
TC08001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
ENST00000378776.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
ENST00000448464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018737.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
TC08001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
ENST00000378776.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
ENST00000448464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018738.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018739.hg.1 |
TC08001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018739.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018739.hg.1 |
ENST00000448464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018739.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018739.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018740.hg.1 |
TC08001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018740.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018740.hg.1 |
ENST00000448464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018740.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018740.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175986.hg.1 |
TC0800011150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175988.hg.1 |
TC0800011150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011150.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175987.hg.1 |
TC0800011150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175989.hg.1 |
TC0800011150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
ENST00000342697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
ENST00000448464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
NM_001135733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
NM_033285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175990.hg.1 |
TC0800011150.hg.1 |
Clariom D TC |