Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001408.2 | NP_001399.1 | 19 - 20 | 6693 | 164 | |
XM_005270580.4 | XP_005270637.1 | 19 - 20 | 7007 | 164 | ||
GenBank mRNA | AF234887.1 | - | 19 - 20 | 6694 | 164 | |
D87469.2 | - | 19 - 20 | 6425 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |