Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004984.2 | NP_004975.2 | 3 - 4 | 499 | 90 | |
GenBank mRNA | AB210045.1 | - | 3 - 4 | 324 | 90 | |
AB290174.1 | - | 3 - 4 | 565 | 90 | ||
AK315202.1 | - | 3 - 4 | 519 | 90 | ||
BC140009.1 | - | 3 - 4 | 552 | 90 | ||
BC146670.1 | - | 3 - 4 | 502 | 40148192 | 90 | |
BC150208.1 | - | 3 - 4 | 502 | 40148890 | 90 | |
U06698.1 | - | 3 - 4 | 439 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |