Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008292.hg.1 |
TC10002945.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008292.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008292.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
ENST00000472998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012491.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186148.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186148.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186148.hg.1 |
ENST00000370489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186148.hg.1 |
NM_020354.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186148.hg.1 |
TC1000012491.hg.1 |
Clariom D TC |