Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000314933.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000432749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000509305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
ENST00000510260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007292.hg.1 |
NM_000491.4 |
Refseq |