Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC032246.1 | - | 11 - 12 | 1453 | 81 |
Gene Symbol: LOC101928574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137602.1 | - | 4 - 5 | 489 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |