Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022383.hg.1 |
TC06001844.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022383.hg.1 |
ENST00000370657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022383.hg.1 |
ENST00000370658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022383.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022383.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022382.hg.1 |
TC06001844.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022382.hg.1 |
ENST00000370657.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022382.hg.1 |
ENST00000370658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022382.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06022382.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012188.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183967.hg.1 |
TC0600012188.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183965.hg.1 |
TC0600012188.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000370657.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000370658.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000485906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
ENST00000622415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
NM_001143940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
NM_016571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600183966.hg.1 |
TC0600012188.hg.1 |
Clariom D TC |