Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002518.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002518.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002518.hg.1 |
TC19000167.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
TC1900006965.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149404.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000495140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
ENST00000497718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006965.hg.1 |
NR_027300.2 |
Refseq |