Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005970.hg.1 |
ENST00000380615.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005970.hg.1 |
ENST00000380617.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005970.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005970.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005970.hg.1 |
TC13000530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000480854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000480854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
ENST00000579551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157305.hg.1 |
TC1300008511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380615.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000380617.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000414289.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000441394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000480854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000480854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
ENST00000579551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_001014380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008511.hg.1 |
NM_032116.4 |
Refseq |