Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017942.hg.1 |
TC09001447.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017942.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017942.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000420475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000497048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011120.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000374672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000411706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000493306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
ENST00000610832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
NM_004235.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175665.hg.1 |
TC0900011120.hg.1 |
Clariom D TC |