Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015175.2 | NP_055990.1 | 24 - 25 | 3717 | 72 | |
XM_006713072.3 | XP_006713135.1 | 23 - 24 | 3633 | 72 | ||
XM_011533533.2 | XP_011531835.1 | 24 - 25 | 3714 | 72 | ||
XM_017006010.1 | XP_016861499.1 | 24 - 25 | 3714 | 72 | ||
XM_017006011.1 | XP_016861500.1 | 24 - 25 | 3570 | 72 | ||
XM_017006012.1 | XP_016861501.1 | 23 - 24 | 3633 | 72 | ||
XM_017006013.1 | XP_016861502.1 | 24 - 25 | 3714 | 72 | ||
XM_017006014.1 | XP_016861503.1 | 23 - 24 | 3489 | 72 | ||
XM_017006015.1 | XP_016861504.1 | 22 - 23 | 3345 | 72 | ||
XM_017006016.1 | XP_016861505.1 | 19 - 20 | 4291 | 72 | ||
GenBank mRNA | AK131104.1 | - | 24 - 25 | 3557 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |