Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 528 | LINC00528 | C22orf37 | -49.7kb | Anti-sense | Chr. 22 - 17777290 - 17779481 on NCBI Build GRCh38 |
Downstream | microRNA 648 | MIR648 |
MIRN648, hsa-mir-648 |
150.1kb | Sense | Chr. 22 - 17980868 - 17980961 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |