Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003046.hg.1 |
ENST00000374518.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003046.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003046.hg.1 |
TC10000266.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150260.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150260.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150260.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150260.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150260.hg.1 |
TC1000007396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
ENST00000374518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007396.hg.1 |
NM_014753.3 |
Refseq |