Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
TC06000023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
ENST00000259808.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
ENST00000541791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000176.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600145572.hg.1 |
TC0600006653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000259808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000380409.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
ENST00000479389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006653.hg.1 |
NM_003804.5 |
Refseq |