Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK024943.1 | - | 1 | 1512 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP binding cassette subfamily C member 8 | ABCC8 |
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2 |
-38.0kb | Sense | Chr. 11 - 17392885 - 17476870 on NCBI Build GRCh38 |
Downstream | otogelin | OTOG |
DFNB18B, MLEMP, OTGN |
31.0kb | Anti-sense | Chr. 11 - 17547373 - 17645944 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |