Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017015634.1 | XP_016871123.1 | 4 - 5 | 2092 | 156 | |
GenBank mRNA | AF435960.1 | - | 1 - 2 | 162 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |