Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
ENST00000360803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
ENST00000538293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
ENST00000545048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000086.hg.1 |
TC10000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000483839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006464.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
ENST00000360803.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
ENST00000491635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
NM_012341.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144253.hg.1 |
TC1000006464.hg.1 |
Clariom D TC |