Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
TC15002755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
ENST00000306901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
ENST00000449991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
ENST00000454250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001129.hg.1 |
NM_001190455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
TC15002755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
ENST00000306901.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
ENST00000449991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
ENST00000454250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001128.hg.1 |
NM_001190455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500146982.hg.1 |
TC1500006766.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000306901.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000437966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
ENST00000454250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NM_000746.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006766.hg.1 |
NR_046324.1 |
Refseq |