Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031667.hg.1 |
TC03001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031667.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031667.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031666.hg.1 |
TC03001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031666.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031666.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191930.hg.1 |
TC0300012283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012283.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191932.hg.1 |
TC0300012283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191933.hg.1 |
TC0300012283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
ENST00000318225.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
ENST00000505070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
NM_152533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
NR_130715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300191931.hg.1 |
TC0300012283.hg.1 |
Clariom D TC |