Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007267.2 | NP_001007268.1 | 18 - 19 | 2875 | 78 | |
NM_001195641.1 | NP_001182570.1 | 18 - 19 | 2875 | 78 | ||
NM_007366.4 | NP_031392.3 | 18 - 19 | 2875 | 78 | ||
XM_005246392.3 | XP_005246449.1 | 18 - 19 | 2763 | 78 | ||
XM_011510820.2 | XP_011509122.1 | 18 - 19 | 2763 | 78 | ||
XM_017003598.1 | XP_016859087.1 | 18 - 19 | 2763 | 78 | ||
XM_017003599.1 | XP_016859088.1 | 18 - 19 | 2763 | 78 | ||
XM_017003600.1 | XP_016859089.1 | 8 - 9 | 986 | 78 | ||
XM_017003602.1 | XP_016859091.1 | 7 - 8 | 712 | 78 | ||
GenBank mRNA | BC140823.1 | - | 18 - 19 | 2685 | 9021691 | 78 |
BC144631.1 | - | 18 - 19 | 2685 | 9053162 | 78 | |
BX485732.1 | - | 6 - 7 | 527 | 78 | ||
D30780.1 | - | 9 - 10 | 1070 | 78 | ||
U17033.1 | - | 18 - 19 | 2875 | 78 | ||
U17034.1 | - | 18 - 19 | 2875 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |