Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005492.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005492.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005492.hg.1 |
TC14000446.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005491.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005491.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005491.hg.1 |
TC14000446.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005489.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005489.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005489.hg.1 |
TC14000446.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005490.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005490.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005490.hg.1 |
TC14000446.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400153821.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153821.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153821.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153821.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153821.hg.1 |
TC1400007649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400153822.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153822.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153822.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153822.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153822.hg.1 |
TC1400007649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007649.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153818.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153818.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153818.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153818.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153818.hg.1 |
TC1400007649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400153819.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153819.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153819.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153819.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153819.hg.1 |
TC1400007649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400153820.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153820.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153820.hg.1 |
ENST00000326303.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400153820.hg.1 |
NM_152331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400153820.hg.1 |
TC1400007649.hg.1 |
Clariom D TC |