Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000262055.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000380123.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000418425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000448283.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000547008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000548516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000550929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
ENST00000552739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NM_001243689.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NM_015416.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NR_045017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NR_045018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NR_045019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
NR_045020.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005054.hg.1 |
TC12000403.hg.1 |
HTA TC |