Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011756.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011756.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011756.hg.1 |
TC22000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011754.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011754.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011754.hg.1 |
TC22000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011755.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011755.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011755.hg.1 |
TC22000662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160940.hg.1 |
TC2200008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160940.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160940.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160940.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160940.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008467.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160938.hg.1 |
TC2200008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160938.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160938.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160938.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160938.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160939.hg.1 |
TC2200008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160939.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160939.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160939.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160939.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160941.hg.1 |
TC2200008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160941.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160941.hg.1 |
NM_015715.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160941.hg.1 |
ENST00000215885.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160941.hg.1 |
NM_015715.4 |
Refseq |