Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
ENST00000355663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
ENST00000388824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
ENST00000402693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013742.hg.1 |
TC19001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
ENST00000355663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
ENST00000388824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
ENST00000402693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013740.hg.1 |
TC19001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013741.hg.1 |
ENST00000388824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013741.hg.1 |
ENST00000402693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013741.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013741.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013741.hg.1 |
TC19001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013743.hg.1 |
ENST00000388824.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013743.hg.1 |
ENST00000402693.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013743.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013743.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013743.hg.1 |
TC19001007.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
TC1900009172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172958.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
TC1900009172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172953.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
TC1900009172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172957.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
TC1900009172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172952.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
NM_203304.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
TC1900009172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
ENST00000402693.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
NM_001174118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172955.hg.1 |
NM_203304.3 |
Refseq |