Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015787.hg.1 |
TC06001324.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015787.hg.1 |
ENST00000378454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015787.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015787.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174784.hg.1 |
TC0600011059.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378450.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000378454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
ENST00000436313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_001195610.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011059.hg.1 |
NM_016356.4 |
Refseq |