Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024936.2 | NP_079212.2 | 10 - 11 | 1247 | 154 | |
XM_011513835.1 | XP_011512137.1 | 11 - 12 | 1259 | 154 | ||
XM_017008128.1 | XP_016863617.1 | 9 - 10 | 1126 | 154 | ||
XM_017008129.1 | XP_016863618.1 | 8 - 9 | 962 | 154 | ||
GenBank mRNA | AK026677.1 | - | 5 - 6 | 516 | 154 | |
AK299211.1 | - | 10 - 11 | 1237 | 154 | ||
AY629351.1 | - | 10 - 11 | 1240 | 154 | ||
BC016914.1 | - | 7 - 8 | 901 | 154 | ||
BC157834.1 | - | 10 - 11 | 1241 | 9057053 | 154 | |
BC171821.1 | - | 10 - 11 | 1241 | 9054475 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |